Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cend1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021316.4 | NP_067291.1 | 2 | 1006 | ||
GenBank mRNA | AF243130.1 | - | 2 | 944 | ||
AK005092.1 | - | 2 | 981 | |||
AK079045.1 | - | 2 | 1007 | |||
BC023032.1 | - | 2 | 944 | 5366891 |