Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290994.1 | NP_001277923.1 | 9 | 5164 | ||
NM_021323.3 | NP_067298.2 | 8 | 5100 | |||
GenBank mRNA | AF229257.1 | - | 9 | 5149 | ||
AK038825.1 | - | 1 | 1349 | |||
AK045405.1 | - | 1 | 340 |