Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163511.1 | NP_001156983.1 | 3 | 396 | ||
NM_021343.2 | NP_067318.2 | 3 | 393 | |||
GenBank mRNA | AF049099.1 | - | 3 | 375 | ||
AK011111.2 | - | 3 | 360 | |||
AK144998.1 | - | 3 | 723 | |||
BC145302.1 | - | 3 | 364 | 9053856 | ||
BC145309.1 | - | 3 | 364 |