Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18180
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
14
3416
NM_001025077.2
NP_001020248.1
13
4613
NM_001083591.1
NP_001077060.1
14
3410
NM_006561.3
NP_006552.3
14
3362
XM_005252354.4
XP_005252411.1
14
3450
XM_005252357.4
XP_005252414.1
11
4409
XM_005252358.3
XP_005252415.1
12
3392
XM_006717373.2
XP_006717436.1
13
3490
XM_011519294.2
XP_011517596.1
14
3432
XM_011519297.1
XP_011517599.1
13
4913
XM_017015542.1
XP_016871031.1
13
3138
XM_017015543.1
XP_016871032.1
13
4586
XM_017015544.1
XP_016871033.1
13
4574
XM_017015545.1
XP_016871034.1
13
4568
XM_017015546.1
XP_016871035.1
14
3210
XM_017015547.1
XP_016871036.1
13
4820
XM_017015548.1
XP_016871037.1
13
4556
XM_017015549.1
XP_016871038.1
14
3198
XM_017015550.1
XP_016871039.1
13
5554
XM_017015551.1
XP_016871040.1
14
3444
XM_017015552.1
XP_016871041.1
13
4943
XM_017015553.1
XP_016871042.1
13
4931
XM_017015554.1
XP_016871043.1
14
3555
XM_017015556.1
XP_016871045.1
12
4860
XM_017015557.1
XP_016871046.1
14
4525
XM_017015558.1
XP_016871047.1
12
4512
XM_017015559.1
XP_016871048.1
12
4848
XM_017015560.1
XP_016871049.1
12
4509
XM_017015561.1
XP_016871050.1
12
4836
XM_017015562.1
XP_016871051.1
12
4830
XM_017015563.1
XP_016871052.1
12
4817
XM_017015564.1
XP_016871053.1
11
4436
XM_017015565.1
XP_016871054.1
11
4439
XM_017015566.1
XP_016871055.1
11
4424
XM_017015567.1
XP_016871056.1
12
4564
XM_017015568.1
XP_016871057.1
14
4551
XM_017015569.1
XP_016871058.1
13
3487
XM_017015570.1
XP_016871059.1
11
4421
XM_017015571.1
XP_016871060.1
12
4833
XM_017015572.1
XP_016871061.1
13
3176
GenBank mRNA
AF036956.1
-
14
3333
AF090693.1
-
13
4590
AF090694.1
-
14
3201
CN356737.1
-
1
24
CR936676.1
-
2
4814
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0