Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021347.4 | NP_067322.1 | 12 | 1534 | ||
GenBank mRNA | AB033595.1 | - | 12 | 1523 | ||
AK028645.1 | - | 12 | 1535 | |||
AK028698.1 | - | 12 | 1535 | |||
AK029062.1 | - | 12 | 1518 | |||
AK080067.1 | - | 1 | 314 |