Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167730.1 | NP_001161202.1 | 11 | 1230 | ||
NM_021385.2 | NP_067360.2 | 10 | 1152 | |||
GenBank mRNA | AF205278.1 | - | 10 | 1076 | ||
AK012795.1 | - | 10 | 1153 | |||
AK028505.1 | - | 10 | 1072 | |||
AK030043.1 | - | 11 | 1231 | |||
BC011120.1 | - | 9 | 1038 | 4014013 |