Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145645.2 | NP_001139117.1 | 1 | 350 | ||
NM_006573.4 | NP_006564.1 | 1 | 350 | |||
GenBank mRNA | AF116456.1 | - | 1 | 201 | ||
AF132600.1 | - | 1 | 219 | |||
AF134715.1 | - | 1 | 227 | |||
AF136293.1 | - | 1 | 83 | |||
AF186114.1 | - | 1 | 240 | |||
AK309629.1 | - | 1 | 251 | |||
AY129225.1 | - | 1 | 83 | |||
AY302751.1 | - | 1 | 83 | |||
AY358881.1 | - | 1 | 252 | |||
BC020674.1 | - | 1 | 262 | 4719315 | ||
BI818459.1 | - | 1 | 350 | |||
BX482686.1 | - | 1 | 295 | |||
CR541818.1 | - | 1 | 83 | |||
HM636064.1 | - | 1 | 83 |