Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021422.4 | NP_067397.1 | 2, 3 | 491 | ||
GenBank mRNA | AB032401.1 | - | 3, 4 | 358 | ||
AK016666.1 | - | 1, 2 | 88 | |||
AK046476.1 | - | 2, 3 | 492 | |||
AK076175.1 | - | 2, 3 | 492 | |||
AK141909.1 | - | 2, 3 | 493 | |||
AK197546.1 | - | 2, 3 | 223 | |||
BC147484.1 | - | 2, 3 | 339 | 9056231 | ||
BC147486.1 | - | 2, 3 | 339 | 9056239 |