Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253832.1 | NP_001240761.1 | 6 | 830 | ||
NM_021438.4 | NP_067413.1 | 6 | 809 | |||
GenBank mRNA | AF270700.1 | - | 6 | 722 | ||
AK008093.1 | - | 6 | 800 | |||
AK008132.1 | - | 6 | 813 | |||
AK012629.1 | - | 6 | 792 | |||
AK019399.1 | - | 6 | 801 | |||
AK028029.1 | - | 6 | 800 | |||
AK086610.1 | - | 3 | 376 | |||
AK207510.1 | - | 2 | 197 | |||
AK208732.1 | - | 2 | 197 | |||
AK210274.1 | - | 2 | 197 | |||
BC031716.1 | - | 6 | 800 | 3969355 | ||
BC049284.1 | - | 6 | 808 | 6395407 |