Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021452.1 | NP_067427.1 | 3 | 966 | ||
GenBank mRNA | AF215892.1 | - | 3 | 966 | ||
AK013647.1 | - | 3 | 1325 | |||
BC048471.1 | - | 3 | 979 |