Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 6 | 945 | ||
NM_001289505.1 | NP_001276434.1 | 8 | 926 | |||
NM_021483.4 | NP_067458.2 | 8 | 1055 | |||
GenBank mRNA | AB032591.1 | - | 8 | 1050 | ||
AK039764.1 | - | 8 | 925 | |||
AK044552.1 | - | 8 | 1183 | |||
AK083141.1 | - | 6 | 837 | |||
HQ646186.1 | - | 9 | 1274 | |||
HQ646187.1 | - | 8 | 1208 | |||
HQ646188.1 | - | 7 | 1103 | |||
HQ646189.1 | - | 7 | 1136 | |||
HQ646190.1 | - | 6 | 1031 | |||
HQ646191.1 | - | 8 | 1864 | |||
HQ646192.1 | - | 9 | 1278 | |||
HQ646193.1 | - | 9 | 1184 | |||
HQ646194.1 | - | 8 | 1079 | |||
HQ646195.1 | - | 9 | 1052 | |||
HQ646196.1 | - | 8 | 947 |