Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 7, 8 | 1092 | ||
NM_001163517.2 | NP_001156989.1 | 2, 3 | 214 | |||
NM_001289505.1 | NP_001276434.1 | 9, 10 | 1073 | |||
NM_021483.4 | NP_067458.2 | 9, 10 | 1202 | |||
GenBank mRNA | AB032591.1 | - | 9, 10 | 1197 | ||
AK006035.1 | - | 2, 3 | 215 | |||
AK039764.1 | - | 9, 10 | 1072 | |||
AK044552.1 | - | 9, 10 | 1330 | |||
AK083141.1 | - | 7, 8 | 984 | |||
HQ646186.1 | - | 10, 11 | 1421 | |||
HQ646187.1 | - | 9, 10 | 1355 | |||
HQ646188.1 | - | 8, 9 | 1250 | |||
HQ646189.1 | - | 8, 9 | 1283 | |||
HQ646190.1 | - | 7, 8 | 1178 | |||
HQ646191.1 | - | 9, 10 | 2011 | |||
HQ646192.1 | - | 10, 11 | 1425 | |||
HQ646193.1 | - | 10, 11 | 1331 | |||
HQ646194.1 | - | 9, 10 | 1226 | |||
HQ646195.1 | - | 10, 11 | 1199 | |||
HQ646196.1 | - | 9, 10 | 1094 | |||
HQ646197.1 | - | 2, 3 | 214 |