Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 11 | 1630 | ||
NM_001163517.2 | NP_001156989.1 | 6 | 752 | |||
NM_001289505.1 | NP_001276434.1 | 13 | 1611 | |||
NM_021483.4 | NP_067458.2 | 13 | 1740 | |||
GenBank mRNA | AB032591.1 | - | 13 | 1735 | ||
AK006035.1 | - | 6 | 753 | |||
AK039764.1 | - | 13 | 1610 | |||
AK083141.1 | - | 11 | 1522 | |||
HQ646186.1 | - | 14 | 1959 | |||
HQ646187.1 | - | 13 | 1893 | |||
HQ646188.1 | - | 12 | 1788 | |||
HQ646189.1 | - | 12 | 1821 | |||
HQ646190.1 | - | 11 | 1716 | |||
HQ646191.1 | - | 13 | 2549 | |||
HQ646192.1 | - | 14 | 1963 | |||
HQ646193.1 | - | 14 | 1869 | |||
HQ646194.1 | - | 13 | 1764 | |||
HQ646195.1 | - | 14 | 1737 | |||
HQ646196.1 | - | 13 | 1632 | |||
HQ646197.1 | - | 6 | 752 |