Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 3 | 534 | ||
NM_001289517.1 | NP_001276446.1 | 3 | 534 | |||
NM_001289518.1 | NP_001276447.1 | 3 | 534 | |||
NM_001289519.1 | NP_001276448.1 | 3 | 534 | |||
NM_001289520.1 | NP_001276449.1 | 3 | 534 | |||
NM_021505.3 | NP_067480.2 | 3 | 534 | |||
GenBank mRNA | AB041593.1 | - | 3 | 477 | ||
AK013978.1 | - | 3 | 458 | |||
AK031432.1 | - | 3 | 459 | |||
AK049719.1 | - | 3 | 488 | |||
AK075969.1 | - | 3 | 473 | |||
AK088327.1 | - | 3 | 497 | |||
AK147951.1 | - | 3 | 473 | |||
AK148679.1 | - | 3 | 489 | |||
AK151314.1 | - | 3 | 480 | |||
AK151905.1 | - | 3 | 480 | |||
AK152528.1 | - | 3 | 450 | |||
AK152981.1 | - | 3 | 480 | |||
AK153373.1 | - | 3 | 480 | |||
AK153422.1 | - | 3 | 480 | |||
AK159204.1 | - | 3 | 480 | |||
AK159713.1 | - | 3 | 473 | |||
AK159916.1 | - | 3 | 473 | |||
AK167561.1 | - | 3 | 473 | |||
AK168135.1 | - | 3 | 458 | |||
AK168737.1 | - | 3 | 474 | |||
AK168814.1 | - | 3 | 473 | |||
AK169196.1 | - | 3 | 497 | |||
AK186196.1 | - | 3 | 212 | |||
AK192754.1 | - | 3 | 230 | |||
AK194108.1 | - | 3 | 199 | |||
AK197886.1 | - | 3 | 206 | |||
AK203216.1 | - | 2 | 183 | |||
AK215079.1 | - | 3 | 207 | |||
AK217317.1 | - | 3 | 212 | |||
BC010339.1 | - | 3 | 450 | 2648173 | ||
BC046566.1 | - | 3 | 513 | 2650790 | ||
BC046804.1 | - | 3 | 471 | 6404923 | ||
BQ950394.1 | - | 3 | 534 |