Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021509.5 | NP_067484.2 | 12 | 2292 | ||
GenBank mRNA | AB041606.1 | - | 12 | 2227 | ||
AK014340.1 | - | 12 | 2293 | |||
AK030274.1 | - | 12 | 2233 | |||
AK162030.1 | - | 12 | 2234 | |||
BC025892.1 | - | 12 | 1990 | |||
BC057652.1 | - | 12 | 2206 | 4017199 |