Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 6 | 720 | ||
GenBank mRNA | AK052983.1 | - | 4 | 3107 | ||
AK088169.1 | - | 6 | 673 | |||
AK161491.1 | - | 818 | ||||
AK165283.1 | - | 1 | 65 | |||
AK178902.1 | - | 3 | 327 | |||
BC016459.1 | - | 4 | 3121 | |||
BC027002.1 | - | 4 | 1572 | |||
BC042187.1 | - | 6 | 655 | 5376052 | ||
BC056224.1 | - | 5 | 650 | 6515270 | ||
BU058854.1 | - | 4 | 473 | |||
Y14196.1 | - | 6 | 678 |