Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 10, 11 | 1235 | ||
GenBank mRNA | AK052983.1 | - | 8, 9 | 3622 | ||
AK088169.1 | - | 10, 11 | 1188 | |||
AK161491.1 | - | 1333 | ||||
AK165283.1 | - | 5, 6 | 580 | |||
BC016459.1 | - | 8, 9 | 3636 | |||
BC027002.1 | - | 8, 9 | 2087 | |||
BC042187.1 | - | 10, 11 | 1170 | 5376052 | ||
BC056224.1 | - | 9, 10 | 1165 | 6515270 | ||
CO428102.1 | - | 2, 3 | 71 | |||
Y14196.1 | - | 10, 11 | 1193 |