Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 14 | 2032 | ||
GenBank mRNA | AK088169.1 | - | 13 | 1935 | ||
AK161491.1 | - | 2130 | ||||
AK165283.1 | - | 9 | 1377 | |||
BC016459.1 | - | 11 | 4383 | |||
BC027002.1 | - | 12 | 2885 | |||
BC042187.1 | - | 14 | 1967 | 5376052 | ||
BC056224.1 | - | 13 | 1962 | 6515270 | ||
Y14196.1 | - | 14 | 1990 |