Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146001.1 | NP_001139473.1 | 3 | 389 | ||
NM_021517.2 | NP_067492.2 | 3 | 435 | |||
GenBank mRNA | AF220100.1 | - | 3 | 374 | ||
AF474247.1 | - | 2 | 373 | |||
AK006269.1 | - | 3 | 909 | |||
AK012070.1 | - | 3 | 425 | |||
AK014879.1 | - | 3 | 692 | |||
AK029752.1 | - | 3 | 893 | |||
AK029764.1 | - | 3 | 903 | |||
AK165415.1 | - | 3 | 370 | |||
BC013512.1 | - | 2 | 358 | 4234949 |