Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 4 | 444 | ||
GenBank mRNA | AB232585.1 | - | 4 | 225 | ||
AK042862.1 | - | 4 | 414 | |||
AK079071.1 | - | 4 | 445 | |||
AK135421.1 | - | 4 | 428 | |||
AK146179.1 | - | 4 | 429 | |||
AK150030.1 | - | 4 | 476 | |||
AK159607.1 | - | 4 | 465 | |||
AK160333.1 | - | 4 | 445 | |||
AK163320.1 | - | 4 | 427 | |||
AK180826.1 | - | 60 | ||||
AK184879.1 | - | 3 | 128 | |||
BC058382.1 | - | 4 | 442 | 6820798 | ||
X95403.1 | - | 4 | 432 |