Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021519.1 | NP_067494.1 | 5 | 638 | ||
GenBank mRNA | AB030185.1 | - | 5 | 638 | ||
AK002345.1 | - | 5 | 653 | |||
AK003557.1 | - | 5 | 665 | |||
AK010191.1 | - | 5 | 653 | |||
AK219893.1 | - | 1 | 113 | |||
BC023472.1 | - | 5 | 632 | 5250117 |