Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021526.2 | NP_067501.2 | 8, 9 | 890 | ||
GenBank mRNA | AK012013.1 | - | 8, 9 | 821 | ||
AK031863.1 | - | 8, 9 | 827 | |||
AK078134.1 | - | 8, 9 | 842 | |||
AK150280.1 | - | 8, 9 | 826 | |||
AK151104.1 | - | 8, 9 | 873 | |||
AK151733.1 | - | 8, 9 | 873 | |||
BC003742.1 | - | 8, 9 | 795 | 3599671 | ||
Y13071.1 | - | 8, 9 | 811 |