Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021531.6 | NP_067506.2 | 4, 5 | 667 | ||
NM_153141.1 | NP_694781.1 | 4, 5 | 667 | |||
GenBank mRNA | AF117887.2 | - | 4, 5 | 577 | ||
AK158242.1 | - | 4, 5 | 633 | |||
AK158757.1 | - | 4, 5 | 507 | |||
BC036974.1 | - | 4, 5 | 651 | 4935077 | ||
BC071267.1 | - | 5, 6 | 783 | |||
BU152197.1 | - | 4, 5 | 667 |