Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021547.3 | NP_067522.1 | 3 | 481 | ||
GenBank mRNA | AK041538.1 | - | 3 | 437 | ||
AK150166.1 | - | 3 | 425 | |||
AK150254.1 | - | 3 | 425 | |||
AK155124.1 | - | 3 | 420 | |||
AK160480.1 | - | 3 | 414 | |||
BC003313.1 | - | 3 | 398 | 3155179 | ||
BC058727.1 | - | 4 | 581 | |||
X82457.1 | - | 3 | 465 |