Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021551.4 | NP_067526.2 | 7 | 1393 | ||
GenBank mRNA | AB012808.1 | - | 9 | 1179 | ||
AK007059.1 | - | 7 | 1338 | |||
AK163116.1 | - | 3 | 2752 | |||
AK196925.1 | - | 2 | 103 | |||
BC005727.1 | - | 6 | 1450 | |||
BC062878.1 | - | 7 | 1412 | 5696414 |