Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021551.4 | NP_067526.2 | 9 | 2166 | ||
GenBank mRNA | AB012808.1 | - | 11 | 1952 | ||
AK007059.1 | - | 9 | 2111 | |||
AK029480.1 | - | 3 | 775 | |||
AK163116.1 | - | 3 | 3939 | |||
AK195316.1 | - | 1 | 247 | |||
BB809257.1 | - | 1 | 254 | |||
BC005727.1 | - | 7 | 2478 | |||
BC062878.1 | - | 9 | 2185 | 5696414 |