Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021551.4 | NP_067526.2 | 9 | 2172 | ||
GenBank mRNA | AB012808.1 | - | 11 | 1958 | ||
AK007059.1 | - | 9 | 2117 | |||
AK029480.1 | - | 3 | 781 | |||
AK163116.1 | - | 3 | 3945 | |||
AK195316.1 | - | 1 | 253 | |||
BB809257.1 | - | 1 | 260 | |||
BC005727.1 | - | 7 | 2484 | |||
BC062878.1 | - | 9 | 2191 | 5696414 |