Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021552.5 | NP_067527.3 | 6 | 927 | ||
GenBank mRNA | AF238866.1 | - | 6 | 853 | ||
AK002474.1 | - | 5 | 778 | |||
AK009136.1 | - | 5 | 856 | |||
AK012832.1 | - | 6 | 868 | |||
AK014275.1 | - | 6 | 904 | |||
AK017605.1 | - | 6 | 868 | |||
AK078457.1 | - | 6 | 880 | |||
BC086776.1 | - | 5 | 723 | |||
BC108365.1 | - | 6 | 788 | 4219428 | ||
BC116384.1 | - | 6 | 868 | |||
BC126976.1 | - | 6 | 867 | 40101726 | ||
BC172160.1 | - | 6 | 839 |