Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021559.2 | NP_067534.2 | 2 | 467 | ||
GenBank mRNA | AB221566.1 | - | 1 | 326 | ||
AF149093.1 | - | 1 | 326 | |||
AK146726.1 | - | 2 | 465 | |||
AY832930.1 | - | 2 | 506 | |||
BC011345.1 | - | 2 | 510 | 3992743 | ||
BC054832.1 | - | 2 | 467 | 6467818 |
Gene Symbol: Zf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084191.1 | - | 1 | 65 |