Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021567.5 | NP_067542.3 | 5 | 422 | ||
GenBank mRNA | AF176328.1 | - | 5 | 308 | ||
AK004580.1 | - | 5 | 373 | |||
AK005383.1 | - | 5 | 371 | |||
AK077632.1 | - | 5 | 372 | |||
AK082005.1 | - | 5 | 372 | |||
AK178757.1 | - | 1 | 41 | |||
AK179721.1 | - | 1 | 41 | |||
AK179793.1 | - | 1 | 41 | |||
AK179802.1 | - | 1 | 41 | |||
AK187694.1 | - | 1 | 41 | |||
AK188206.1 | - | 1 | 41 | |||
AK188263.1 | - | 1 | 41 | |||
AK189619.1 | - | 1 | 41 | |||
BC010694.1 | - | 5 | 311 | 2654291 | ||
BY259572.1 | - | 5 | 409 | |||
CJ162439.1 | - | 5 | 419 | |||
CJ174803.1 | - | 5 | 417 |