Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021605.3 | NP_067618.1 | 7 | 672 | ||
GenBank mRNA | AF217650.1 | - | 6 | 523 | ||
AK035502.1 | - | 7 | 671 | |||
AK078639.1 | - | 7 | 681 | |||
AK088173.1 | - | 7 | 721 | |||
AK149308.1 | - | 7 | 720 | |||
AK165583.1 | - | 7 | 650 | |||
BC026541.1 | - | 7 | 664 | |||
BC037697.1 | - | 7 | 649 | 3994871 |