Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC125528.1 | - | 4 | 430 | ||
BY714594.1 | - | 1 | 117 | |||
CJ236701.1 | - | 4 | 247 | |||
X65979.1 | - | 3 | 255 | |||
X65981.1 | - | 5 | 552 |