Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162858.1 | NP_001156330.1 | 1, 2 | 547 | ||
NM_021717.2 | NP_068363.2 | 1, 2 | 547 | |||
GenBank mRNA | AJ278170.1 | - | 1, 2 | 570 | ||
AK134689.1 | - | 1, 2 | 344 | |||
AK156526.1 | - | 1, 2 | 420 | |||
BC094230.1 | - | 1, 2 | 265 | 30535040 | ||
BY718516.1 | - | 1, 2 | 342 |