Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHCYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242673.1 | NP_001229602.1 | 3 | 968 | ||
NM_001242674.1 | NP_001229603.1 | 4 | 338 | |||
NM_001242675.1 | NP_001229604.1 | 3 | 692 | |||
NM_001242676.1 | NP_001229605.1 | 3 | 681 | |||
NM_006621.5 | NP_006612.2 | 3 | 597 | |||
XM_011540535.1 | XP_011538837.1 | 3 | 620 | |||
GenBank mRNA | AF315687.1 | - | 3 | 676 | ||
AK303690.1 | - | 3 | 244 | |||
AK308080.1 | - | 3 | 303 | |||
AK316110.1 | - | 3 | 496 | |||
AL049954.1 | - | 3 | 511 | |||
BC007576.1 | - | 3 | 525 | 3139729 | ||
BC010681.1 | - | 3 | 584 | 3853747 | ||
BC016942.1 | - | 3 | 555 | 3450568 | ||
BC065254.1 | - | 3 | 588 | 6138596 | ||
BC095476.1 | - | 3 | 540 | 6179838 | ||
BC110896.1 | - | 3 | 515 | 5768161 | ||
BC111563.1 | - | 3 | 234 | |||
U82761.1 | - | 3 | 265 |