Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021887.2 | NP_068687.1 | 9 | 1345 | ||
GenBank mRNA | AB049137.1 | - | 9 | 1237 | ||
AF254068.1 | - | 9 | 1049 | |||
AF269134.1 | - | 8 | 907 | |||
AF279436.1 | - | 9 | 1297 | |||
AF477982.1 | - | 8 | 923 | |||
AF477983.1 | - | 8 | 923 | |||
AF477984.1 | - | 8 | 923 | |||
AF477985.1 | - | 8 | 923 | |||
AF477986.1 | - | 8 | 923 | |||
AK150824.1 | - | 9 | 1306 | |||
AK171826.1 | - | 9 | 1284 | |||
BC057861.1 | - | 8 | 1197 | 5315350 |