Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 1 | 322 | ||
XM_011541226.1 | XP_011539528.1 | 1 | 381 | |||
XM_011541231.2 | XP_011539533.1 | 1 | 183 | |||
GenBank mRNA | AF171237.1 | - | 1 | 168 | ||
AK309662.1 | - | 1 | 182 | |||
AK310132.1 | - | 1 | 182 | |||
AK315360.1 | - | 1 | 243 | |||
BC000303.2 | - | 1 | 162 | 2820369 | ||
BC001349.2 | - | 1 | 157 | 2961389 | ||
BC011262.1 | - | 1 | 231 | 4156703 | ||
BC023235.1 | - | 1 | 135 | |||
CR456795.1 | - | 1 | 95 | |||
DA731339.1 | - | 1 | 403 | |||
FJ224347.1 | - | 787 |