Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021889.4 | NP_068689.2 | 2 | 512 | ||
GenBank mRNA | AB026802.1 | - | 2 | 311 | ||
AK089115.1 | - | 2 | 513 | |||
AK156631.1 | - | 2 | 513 | |||
BC132495.1 | - | 2 | 499 | 40130772 | ||
BC137904.1 | - | 2 | 499 | 8860923 | ||
BC145024.1 | - | 2 | 499 |