Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND11 SIRNA Assays ›
Gene Symbol:
ZMYND11
Gene Name:
zinc finger MYND-type containing 11
Gene Aliases:
BRAM1, BS69, MRD30
Chromosome Location:
Chr.10: 134340 - 254637 on Build GRCh38
UniGene ID:
Hs.292265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18217
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202464.1
NP_001189393.1
3
326
NM_001202465.1
NP_001189394.1
3
659
NM_001202466.1
NP_001189395.1
3
659
NM_001202467.1
NP_001189396.1
2
250
NM_001202468.1
NP_001189397.1
2
250
NM_006624.5
NP_006615.2
3
572
NM_212479.3
NP_997644.2
3
659
XM_005252359.4
XP_005252416.1
3
412
XM_005252361.3
XP_005252418.1
3
495
XM_005252362.2
XP_005252419.1
3
659
XM_006717376.2
XP_006717439.1
3
697
XM_017015587.1
XP_016871076.1
3
325
XM_017015588.1
XP_016871077.1
3
561
XM_017015589.1
XP_016871078.1
3
402
XM_017015590.1
XP_016871079.1
3
859
XM_017015591.1
XP_016871080.1
4
372
XM_017015592.1
XP_016871081.1
3
402
XM_017015593.1
XP_016871082.1
3
842
XM_017015594.1
XP_016871083.1
3
330
GenBank mRNA
AK294469.1
-
3
341
AK294781.1
-
3
341
AK296427.1
-
2
222
AK296518.1
-
1
53
AK312570.1
-
3
309
AY732489.1
-
1
111
BC012586.2
-
2
251
BC034784.1
-
3
440
5171494
BC091489.1
-
3
326
6186814
DB236591.1
-
3
432
DQ335452.1
-
2
231
DQ335453.1
-
2
231
DQ335454.1
-
2
231
DQ335455.1
-
2
231
X86098.1
-
3
355
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_144516, NM_001199141, BC058999, CT010329, CB526775, and others.
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
cell proliferation
viral process
covalent chromatin modification
regulation of transcription elongation from RNA polymerase II promoter
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of JNK cascade
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
transcription corepressor activity
protein binding
zinc ion binding
methylated histone binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0