Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021895.2 | NP_068695.1 | 15 | 1813 | ||
GenBank mRNA | AJ289242.2 | - | 15 | 1813 | ||
AK030237.1 | - | 8 | 983 | |||
AK145322.1 | - | 15 | 1866 | |||
AK150044.1 | - | 15 | 1815 | |||
BC013616.1 | - | 15 | 1822 | 3157024 | ||
BC027032.1 | - | 15 | 1762 | |||
BC087554.1 | - | 15 | 1766 | 5351470 | ||
DQ303411.1 | - | 15 | 1716 |