Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021896.5 | NP_068696.2 | 2 | 536 | ||
GenBank mRNA | AF297082.1 | - | 2 | 511 | ||
AK004815.1 | - | 2 | 470 | |||
AK028558.1 | - | 2 | 459 | |||
AK031305.1 | - | 2 | 449 | |||
BC057327.1 | - | 2 | 446 | 6825960 | ||
BC094310.1 | - | 2 | 592 | 6838836 |