Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021896.5 | NP_068696.2 | 5, 6 | 1403 | ||
GenBank mRNA | AF297082.1 | - | 5, 6 | 1378 | ||
AK004815.1 | - | 5, 6 | 1337 | |||
AK028558.1 | - | 5, 6 | 1326 | |||
BC057327.1 | - | 5, 6 | 1313 | 6825960 | ||
BC094310.1 | - | 5, 6 | 1459 | 6838836 |