Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 4 | 778 | ||
GenBank mRNA | AB021875.2 | - | 3 | 705 | ||
AF208026.1 | - | 4 | 746 | |||
AF263522.1 | - | 3 | 736 | |||
AF279672.1 | - | 4 | 762 | |||
AJ296078.1 | - | 3 | 697 | |||
AK002735.1 | - | 4 | 779 | |||
AK083223.1 | - | 4 | 748 | |||
BC127052.1 | - | 4 | 743 | 40109678 |