Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022022.3 | NP_071305.2 | 18, 19 | 2939 | ||
GenBank mRNA | AF260926.1 | - | 18, 19 | 3054 | ||
AK029914.1 | - | 18, 19 | 2544 | |||
AK083409.1 | - | 18, 19 | 2542 | |||
AK122345.1 | - | 18, 19 | 2450 | |||
AK163319.1 | - | 6, 7 | 735 | |||
BC067402.1 | - | 18, 19 | 2686 | 30099547 | ||
BC075620.1 | - | 18, 19 | 2900 | 6405514 |