Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098486.1 | NP_001091956.1 | 5, 6 | 743 | ||
NM_006632.3 | NP_006623.2 | 4, 5 | 509 | |||
GenBank mRNA | AK296070.1 | - | 3, 4 | 467 | ||
AK296082.1 | - | 4, 5 | 699 | |||
AK298271.1 | - | 5, 6 | 743 | |||
AK298367.1 | - | 4, 5 | 506 | |||
BC017952.1 | - | 4, 5 | 478 | 4557795 | ||
U90545.1 | - | 4, 5 | 718 | |||
Z83953.1 | - | 2, 3 | 332 |