Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006636.3 | NP_006627.2 | 2 | 215 | ||
NR_027405.1 | 3 | 313 | ||||
GenBank mRNA | AK130664.1 | - | 2 | 163 | ||
AK222826.1 | - | 2 | 165 | |||
AK223041.1 | - | 2 | 163 | |||
AK313138.1 | - | 2 | 163 | |||
AU125762.1 | - | 2 | 194 | |||
BC001548.1 | - | 2 | 150 | 3462450 | ||
BC015062.1 | - | 3 | 263 | 4285669 | ||
BC017054.1 | - | 2 | 143 | 3846710 | ||
BX475613.1 | - | 2 | 216 | |||
X16396.1 | - | 2 | 133 |