Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162500.1 | NP_001155972.1 | 13 | 1093 | ||
NM_022321.4 | NP_071716.3 | 13 | 1048 | |||
GenBank mRNA | AF312712.1 | - | 13 | 1006 | ||
AK028329.1 | - | 13 | 1049 | |||
AK037365.1 | - | 13 | 1021 | |||
AK040914.1 | - | 13 | 979 | |||
AK079881.1 | - | 13 | 974 | |||
AK088419.1 | - | 13 | 998 | |||
BC011200.1 | - | 13 | 1002 | 4190510 |