Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 6 | 1291 | ||
GenBank mRNA | AF136277.1 | - | 6 | 1301 | ||
AK002710.1 | - | 6 | 1253 | |||
AK004095.1 | - | 6 | 1289 | |||
AK008370.1 | - | 6 | 1220 | |||
AK010912.1 | - | 6 | 1287 | |||
AK150329.1 | - | 6 | 1219 | |||
AK150894.1 | - | 6 | 1226 | |||
AK150947.1 | - | 6 | 1280 | |||
AK151585.1 | - | 6 | 1254 | |||
AK151994.1 | - | 6 | 1264 | |||
AK153147.1 | - | 6 | 1226 | |||
AK159118.1 | - | 6 | 1237 | |||
AK159618.1 | - | 6 | 1304 | |||
AK159826.1 | - | 6 | 1224 | |||
AK160001.1 | - | 6 | 1274 | |||
AK160028.1 | - | 6 | 1274 | |||
AK160354.1 | - | 6 | 1307 | |||
AK166047.1 | - | 6 | 1275 | |||
AK170963.1 | - | 6 | 1274 | |||
AK218117.1 | - | 1 | 20 | |||
AK219014.1 | - | 1 | 20 | |||
BC008619.1 | - | 6 | 1291 | 3588457 |