Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022327.5 | NP_071722.1 | 5 | 1161 | ||
GenBank mRNA | AF174296.1 | - | 1129 | |||
AK017698.1 | - | 5 | 1007 | |||
AK033193.1 | - | 5 | 1016 | |||
AK159993.1 | - | 5 | 1106 | |||
BC006907.1 | - | 5 | 1005 | 3599947 |