Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022378.3 | NP_071773.2 | 2 | 956 | ||
GenBank mRNA | BC111908.1 | - | 2 | 667 | 40045942 | |
U90538.1 | - | 2 | 903 | |||
X92592.1 | - | 2 | 841 |