Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022413.4 | NP_071858.3 | 4, 5 | 777 | ||
GenBank mRNA | AJ542487.1 | - | 4, 5 | 482 | ||
AK077234.1 | - | 4, 5 | 707 | |||
AK168760.1 | - | 4, 5 | 777 | |||
BC016101.1 | - | 4, 5 | 715 | 4911355 |